3-Fluorophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A659930-1G A659930-5G
103840-438EA 19 USD
103840-438 76844-628 76844-630
3-Fluorophenylboronic acid pinacol ester 97%
3-Fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BFO₂
Melting Pt: 212…215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18375236
CAS Number: 936618-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR