2-(3-Chlorophenyl)ethanol 97%
Supplier: Ambeed
|
Warning
|
A229034-25G
A229034-5G
A229034-1G
103819-440EA
6
USD
103819-440
103850-340
77089-160
77089-162
77303-842
2-(3-Chlorophenyl)ethanol 97%
2-(3-Chlorophenyl)ethanol
Precaución: for Research Use Only
Formula:
C₈H₉ClO Boiling Pt: 257.2 °C (760 mm Hg) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002892 CAS Number: 5182-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...