3-Chlorophenylboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(3-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Chlorophenylboronic acid, pinacol ester
A128456-100G
A128456-10G
A128456-1G
A128456-25G
A128456-5G
103801-126EA
6
USD
103801-126
77271-888
77271-890
77313-432
77313-434
77313-436
3-Chlorophenylboronic acid pinacol ester 97%
3-Chlorophenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BClO₂ Boiling Pt: 317.893 °C (760 mm Hg) Flash Pt: 146.057 °C Storage Temperature: Ambient |
MDL Number:
MFCD05663877 CAS Number: 635305-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...