(3-Chloro-2-fluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Chloro-2-fluorophenyl)acetic acid

A172300-1G A172300-100G
103806-712EA 7 USD
103806-712 103844-996 77082-188 77273-760
(3-Chloro-2-fluorophenyl)acetic acid 97%
(3-Chloro-2-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃ClF)CH₂COOH
Boiling Pt: 296.4 °C (760 mm Hg)
Flash Pt: 133.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01631554
CAS Number: 261762-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR