2-(3-But-3-ynyl-3H-diazirin-3-yl)-ethanol 98%

Supplier: Ambeed
Danger

A118147-100MG A118147-250MG A118147-50MG A118147-1G
103792-060EA 174 USD
103792-060 103799-002 77233-966 77233-968 77233-970 77297-584
2-(3-But-3-ynyl-3H-diazirin-3-yl)-ethanol 98%
2-(3-But-3-ynyl-3H-diazirin-3-yl)-ethanol

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
Boiling Pt: 204.1±36.0 °C (760 mm Hg)
Flash Pt: 106.7 °C
Storage Temperature: Freezer
MDL Number: MFCD29918290
CAS Number: 1450754-41-2
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR