2-(3-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromophenylboronic acid Pinacol ester, 1-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
A168302-100G
A168302-250MG
A168302-10G
A168302-25G
103806-272EA
6
USD
103806-272
103844-528
76813-116
76813-118
77593-030
77593-032
2-(3-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(3-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BBrO₂ Boiling Pt: 333.2±25.0 °C (760 mm Hg) Flash Pt: 155.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12545926 CAS Number: 594823-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...