3-Bromobenzaldehyde ethylene acetal 97% (by GC)

Supplier: Ambeed
Warning

A154172-100G A154172-10G A154172-1G A154172-25G A154172-5G
103804-610EA 5 USD
103804-610 103842-790 77259-386 77259-388 77300-042 77318-484 77318-486
3-Bromobenzaldehyde ethylene acetal 97% (by GC)
3-Bromobenzaldehyde ethylene acetal

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Boiling Pt: 278.5 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003209
CAS Number: 17789-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR