3-Benzoylphenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Benzoylphenyl)acetic acid, Desmethyl Ketoprofen

A121140-250MG A121140-1G A121140-5G A121140-100MG
103800-032EA 213 USD
103800-032 76781-756 77312-152 77312-154 77871-535
3-Benzoylphenylacetic acid 97%
3-Benzoylphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₃
Boiling Pt: 438.5 °C (760 mm Hg)
Flash Pt: 233.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00046549
CAS Number: 22071-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR