(3,5-Dimethylisoxazol-4-yl)acetic acid 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A226330-5G A226330-100MG A226330-250MG A226330-25G
103819-238EA 83 USD
103819-238 103850-140 77088-868 77165-394 77683-656 77892-361
(3,5-Dimethylisoxazol-4-yl)acetic acid 98%, Ambeed.Inc
(3,5-Dimethylisoxazol-4-yl)acetic acid

Precaución: For research use only.

Formula: C₇H₉NO₃
Boiling Pt: 322.5 °C (760 mm Hg)
Melting Pt: 120…121 °C at N/A mmHg
Flash Pt: 148.8 °C
Storage Temperature: Ambient
MDL Number: MFCD05864546
CAS Number: 2510-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR