(3,5-Dimethoxyphenyl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethoxyphenylacetonitrile, 3,5-Dimethoxybenzyl cyanide, 2-(3,5-Dimethoxyphenyl)acetonitrile

A179108-10G A179108-250MG A179108-1G A179108-5G A179108-100G
103807-394EA 10 USD
103807-394 103845-734 76682-480 76682-482 77083-182 77301-592 77598-236
(3,5-Dimethoxyphenyl)acetonitrile 97%
(3,5-Dimethoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 171…178 °C (760 mm Hg)
Melting Pt: 54…57 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00016395
CAS Number: 13388-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR