(3,5-Dimethoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Dimethoxyphenyl)acetic acid

A177039-100G A177039-10G A177039-250MG A177039-1G A177039-25G A177039-5G
103807-204EA 16 USD
103807-204 103845-530 77273-900 77273-902 77273-904 77301-486 77301-488 77301-490
(3,5-Dimethoxyphenyl)acetic acid 97%
(3,5-Dimethoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196.2 g/mol
Boiling Pt: 352.6 °C (760 mm Hg)
Melting Pt: 102…103 °C
Flash Pt: 140.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00016827
CAS Number: 4670-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR