(3,5-Dichlorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Dichlorophenyl)acetic acid

A101831-1000G A101831-100G A101831-10G A101831-1G
103796-164EA 5 USD
103796-164 103821-856 77233-098 77233-100 77233-102 77282-842
(3,5-Dichlorophenyl)acetic acid 97%
(3,5-Dichlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MW: 205.03 g/mol
Flash Pt: 150.5 °C
Storage Temperature: Freezer
MDL Number: MFCD01764671
CAS Number: 51719-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR