3,5-Dibromophenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Dibromophenyl)acetic acid

A212573-1G A212573-5G A212573-10G
103818-214EA 9 USD
103818-214 103848-910 77087-374 77087-376 77275-222
3,5-Dibromophenylacetic acid 98%
3,5-Dibromophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Boiling Pt: 381.7 °C (760 mm Hg)
Flash Pt: 184.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07787513
CAS Number: 188347-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR