2-[3,5-Bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
Supplier: Ambeed
|
Warning
|
A130067-1G
A130067-10G
103801-372EA
6
USD
103801-372
77313-804
77885-138
2-[3,5-Bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-[3,5-Bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₄H₁₅BF₆O₂ Boiling Pt: 293.5 °C (760 mm Hg) Flash Pt: 131.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD12405516 CAS Number: 69807-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...