2-(3,4-Dimethoxyphenyl)ethanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Homoveratryl Alcohol, 3,4-Dimethoxyphenethyl alcohol, 3,4-Dimethoxybenzeneethanol, 3,4-Dimethoxyphenethyl Alcohol
A241267-100G
A241267-10G
A241267-25G
A241267-5G
A241267-1G
103811-402EA
5
USD
103811-402
103851-206
76692-284
76692-286
77090-368
77090-370
77304-210
2-(3,4-Dimethoxyphenyl)ethanol 98%
2-(3,4-Dimethoxyphenyl)ethanol
Precaución: for Research Use Only
Formula:
C₁₀H₁₄O₃ MW: 182.22 g/mol Boiling Pt: 295.2 °C (760 mm Hg) Melting Pt: 44…48 °C Flash Pt: 127.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002894 CAS Number: 7417-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...