3,4-Dimethoxyphenylboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,4-Dimethoxyphenylboronic acid Pinacol ester, 1,2-Dimethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-(3,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
A193794-250MG
A193794-1G
A193794-5G
A193794-10G
103816-582EA
6
USD
103816-582
103847-194
76785-038
77085-096
77085-098
77567-806
3,4-Dimethoxyphenylboronic acid pinacol ester 97%
3,4-Dimethoxyphenylboronic acid pinacol ester
Precaución: For research use only.
Formula:
C₁₄H₂₁BO₄ MW: 264.13 g/mol Boiling Pt: 352.5 °C (760 mm Hg) Melting Pt: 86…87 °C Flash Pt: 167 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05863909 CAS Number: 365564-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...