3-Hydroxytyrosol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-Dihydroxyphenyl)ethanol, Hydroxytyrosol, DOPET, Homoprotocatechuyl alcohol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-Dihydroxyphenethyl Alcohol

A172108-1G A172108-25G A172108-100MG A172108-250MG A172108-5G A172108-100G
103792-286EA 14 USD
103792-286 103799-422 77191-450 77191-452 77615-436 77615-438 77615-440 77746-138
3-Hydroxytyrosol 98%
3-Hydroxytyrosol

Precaución: Sealed in dry, store in freezer, under –20 °C.

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01320529
CAS Number: 10597-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR