2-(3,4-Dihydroxyphenyl)-3,7-dihydroxy-4H-chromen-4-one hydrate 97%

Supplier: Ambeed
Warning

A115911-250MG A115911-1G
103798-646EA 47 USD
103798-646 103824-510 77161-000 77311-332
2-(3,4-Dihydroxyphenyl)-3,7-dihydroxy-4H-chromen-4-one hydrate 97%
2-(3,4-Dihydroxyphenyl)-3,7-dihydroxy-4H-chromen-4-one hydrate

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₇
Storage Temperature: Ambient
MDL Number: MFCD04220857
CAS Number: 1174865-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR