2-(3-(Methoxycarbonyl)phenyl)acetic acid 95%

Supplier: Ambeed
Warning

A243218-1G A243218-100MG A243218-250MG
103811-510EA 64 USD
103811-510 103851-334 77090-530 77113-286 77113-288
2-(3-(Methoxycarbonyl)phenyl)acetic acid 95%
2-(3-(Methoxycarbonyl)phenyl)acetic acid
Formula: C₁₀H₁₀O₄
Boiling Pt: 350.8 °C (760 mm Hg)
Melting Pt: 83…84 °C
Flash Pt: 140.6 °C
Storage Temperature: Ambient
MDL Number: MFCD18459189
CAS Number: 52787-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR