2-(2-Phenylacetamido)pentanedioic acid 97%

Supplier: Ambeed
Warning

A101321-100MG A101321-250MG A101321-1G
103795-908EA 45 USD
103795-908 103821-594 77188-552 77188-554 77297-056
2-(2-Phenylacetamido)pentanedioic acid 97%
2-(2-Phenylacetamido)pentanedioic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₅
MW: 265.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00437417
CAS Number: 2752-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR