Phenaceturic acid 97%

Supplier: Ambeed
Warning

A118001-100G A118001-10G A118001-250MG A118001-25G A118001-1G A118001-5G
103798-974EA 6 USD
103798-974 103824-866 76718-382 76718-384 76718-386 76718-388 77311-650 77311-652
Phenaceturic acid 97%
Phenaceturic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
Boiling Pt: 476.5 °C (760 mm Hg)
Melting Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00021744
CAS Number: 500-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR