2-(2-Nitrophenyl)ethanol 98%

Supplier: Ambeed
Warning

A266922-1G A266922-5G A266922-10G
103812-820EA 7 USD
103812-820 103852-722 77092-434 77092-436 77276-694
2-(2-Nitrophenyl)ethanol 98%
2-(2-Nitrophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.17 g/mol
Boiling Pt: 267 °C (760 mm Hg)
Melting Pt: 2 °C
Flash Pt: 129.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00007192
CAS Number: 15121-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR