(2-Nitrophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A114086-100G A114086-10G A114086-25G A114086-500G
103824-186EA 5 USD
103824-186 77271-114 77271-116 77271-118 77271-120
(2-Nitrophenyl)acetic acid 98%
(2-Nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄NO₂)CH₂COOH
Boiling Pt: 345.1 °C (760 mm Hg)
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00007190
CAS Number: 3740-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR