2-(2-Nitro-1H-imidazol-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

A116634-100MG A116634-250MG A116634-1G A116634-5G
103798-774EA 283 USD
103798-774 103824-638 77161-050 77161-052 77311-434 77311-436
2-(2-Nitro-1H-imidazol-1-yl)acetic acid 95%
2-(2-Nitro-1H-imidazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₅N₃O₄
MW: 171.11 g/mol
Boiling Pt: 487.2 °C (760 mm Hg)
Flash Pt: 248.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12022607
CAS Number: 22813-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR