2-(2-Methyl-6-nitrophenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
A372688-1G
A372688-250MG
A372688-100MG
103826-510EA
90
USD
103826-510
103837-528
76840-788
77016-014
77684-079
2-(2-Methyl-6-nitrophenyl)acetic acid 98%
2-(2-Methyl-6-nitrophenyl)acetic acid
Precaución: For research use only.
Formula:
C₉H₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00066313 CAS Number: 23876-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...