2-(2-Methoxypyridin-4-yl)acetic acid 97%

Supplier: Ambeed
Warning

A277869-5G A277869-1G A277869-100MG A277869-250MG
103813-374EA 246 USD
103813-374 76827-448 77093-212 77885-741 77885-742
2-(2-Methoxypyridin-4-yl)acetic acid 97%
2-(2-Methoxypyridin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD10698166
CAS Number: 464152-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR