2-(2-Methoxyphenoxy)ethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A100917-10G A100917-1G A100917-100G A100917-25G A100917-500G
103795-694EA 6 USD
103795-694 103821-384 77245-084 77282-602 77614-732 77745-768 77745-769
2-(2-Methoxyphenoxy)ethanamine hydrochloride 97%
2-(2-Methoxyphenoxy)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNO₂
Boiling Pt: 261.1 °C (760 mm Hg)
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD01631120
CAS Number: 64464-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR