m-PEG2-amine 98%

Supplier: Ambeed
Danger

A188529-100MG A188529-250MG A188529-1G A188529-5G A188529-25G
103816-114EA 17 USD
103816-114 103846-710 76974-760 76974-762 77084-464 77084-466 77671-672
m-PEG2-amine 98%
m-PEG2-amine

Precaución: for Research Use Only

Formula: C₅H₁₃NO₂
Flash Pt: 54.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09032941
CAS Number: 31576-51-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR