(2-Methoxyethoxy)acetic acid 90%

Supplier: Ambeed
Warning

A851755-1G A851755-25G A851755-5G A851755-100G A851755-10G A851755-500G
103841-240EA 11 USD
103841-240 76845-490 76845-492 76845-494 77105-926 77105-928 77105-930
(2-Methoxyethoxy)acetic acid 90%
(2-Methoxyethoxy)acetic acid

Precaución: for Research Use Only

Formula: C₅H₁₀O₄
Boiling Pt: 251.3 °C (760 mm Hg)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00044097
CAS Number: 16024-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR