2-(2-Isopropyloxazolidin-3-yl)ethanol 97%

Supplier: Ambeed
Warning

A101115-25G A101115-1G A101115-5G
103795-806EA 12 USD
103795-806 103821-496 77135-476 77282-662 77296-988
2-(2-Isopropyloxazolidin-3-yl)ethanol 97%
2-(2-Isopropyloxazolidin-3-yl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO₂
Boiling Pt: 241.8 °C (760 mm Hg)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD18818644
CAS Number: 28770-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR