2-(2-Iodophenyl)ethan-1-ol 95%

Supplier: Ambeed
Warning

A146240-250MG A146240-1G A146240-5G
103803-648EA 86 USD
103803-648 103841-734 77010-546 77299-508 77299-510
2-(2-Iodophenyl)ethan-1-ol 95%
2-(2-Iodophenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₉IO
MW: 248.06 g/mol
Boiling Pt: 294.7 °C (760 mm Hg)
Flash Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12031769
CAS Number: 26059-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR