(2-Iodophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Iodophenyl)acetic acid

A183177-1G A183177-25G A183177-5G A183177-100G A183177-10G A183177-250MG
103815-614EA 6 USD
103815-614 103846-176 77083-768 77083-770 77083-772 77236-630 77236-632 77236-634
(2-Iodophenyl)acetic acid 97%
(2-Iodophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄I)CH₂COOH
MW: 262.05 g/mol
Boiling Pt: 344.5 °C (760 mm Hg)
Melting Pt: 116…119 °C
Flash Pt: 162.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00046546
CAS Number: 18698-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR