2-(2-Hydroxyphenyl)ethanol 97%

Supplier: Ambeed
Warning

A167151-1G A167151-25G A167151-5G A167151-100G A167151-10G
103806-138EA 19 USD
103806-138 103844-400 77081-396 77081-398 77081-400 77110-870 77110-872
2-(2-Hydroxyphenyl)ethanol 97%
2-(2-Hydroxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
Boiling Pt: 168…169 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002890
CAS Number: 7768-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR