Furan-2-boronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2-Furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Furanboronic acid pinacol ester
A176072-1G
A176072-5G
103807-114EA
11
USD
103807-114
103845-430
77082-784
77082-786
Furan-2-boronic acid pinacol ester 98%
Furan-2-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₀H₁₅BO₃ Boiling Pt: 255 °C (760 mm Hg) Flash Pt: 108 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02094008 CAS Number: 374790-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...