2-(2-Fluorophenyl)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluorophenethyl alcohol

A417574-1G A417574-25G A417574-5G A417574-10G
103827-334EA 9 USD
103827-334 103838-380 76841-966 76841-968 76841-970 77264-674
2-(2-Fluorophenyl)ethanol 98%
2-(2-Fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 207.8 °C (760 mm Hg)
Flash Pt: 98.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00002887
CAS Number: 50919-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR