2-Chlorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chlorophenylboronic acid Pinacol ester, 2-(2-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

A175068-1G A175068-25G A175068-5G A175068-10G A175068-250MG
103806-996EA 6 USD
103806-996 103845-298 77082-614 77082-616 77082-618 77260-060 77260-062
2-Chlorophenylboronic acid pinacol ester 98%
2-Chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BClO₂
Boiling Pt: 314.9 °C (760 mm Hg)
Flash Pt: 144.3 °C
Storage Temperature: Ambient
MDL Number: MFCD05663846
CAS Number: 870195-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR