(2-Chloro-4-fluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Chloro-4-fluorophenyl)acetic acid

A101899-100G A101899-10G A101899-25G A101899-5G
103821-896EA 13 USD
103821-896 77233-104 77233-106 77233-108 77308-784
(2-Chloro-4-fluorophenyl)acetic acid 98%
(2-Chloro-4-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59 g/mol
Boiling Pt: 292.8 °C (760 mm Hg)
Melting Pt: 101…104 °C
Flash Pt: 130.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00236028
CAS Number: 177985-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR