2-(2-Chloro-3-fluorophenyl)acetic acid 95%
Supplier: Ambeed
|
Warning
|
A279491-250MG
A279491-1G
A279491-5G
A279491-10G
103813-468EA
20
USD
103813-468
103853-414
77032-138
77093-350
77093-352
77872-340
2-(2-Chloro-3-fluorophenyl)acetic acid 95%
2-(2-Chloro-3-fluorophenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₈H₆ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09925136 CAS Number: 1000523-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...