2-(2-Carboxyethyl)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
A111503-250MG
A111503-1G
A111503-5G
A111503-100MG
103797-850EA
29
USD
103797-850
103823-670
77270-934
77310-474
77310-476
77747-822
2-(2-Carboxyethyl)benzoic acid 98%
2-(2-Carboxyethyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O₄ Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016550 CAS Number: 776-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...