2-(2-Bromopyridin-3-yl)acetonitrile 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A100660-100MG A100660-250MG A100660-1G A100660-5G
103795-556EA 298 USD
103795-556 77160-168 77160-170 77296-794 77682-932
2-(2-Bromopyridin-3-yl)acetonitrile 95%, Ambeed.Inc
2-(2-Bromopyridin-3-yl)acetonitrile

Precaución: For research use only.

Formula: C₇H₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD10000072
CAS Number: 1211523-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR