2-(2-Bromophenyl)ethanol 98%
Supplier: Ambeed
|
Warning
|
A161730-100G
A161730-10G
A161730-500G
A161730-1G
A161730-25G
A161730-5G
103805-528EA
9
USD
103805-528
103843-770
77259-626
77259-628
77259-630
77300-554
77319-770
77319-772
2-(2-Bromophenyl)ethanol 98%
2-(2-Bromophenyl)ethanol
Precaución: for Research Use Only
Formula:
C₈H₉BrO Boiling Pt: 269.3 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093566 CAS Number: 1074-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...