(2-Bromophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A259569-25G A259569-100G A259569-10G A259569-500G A259569-5G
103852-332EA 5 USD
103852-332 77091-900 77238-476 77238-478 77238-480 77304-774
(2-Bromophenyl)acetic acid 98%
(2-Bromophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄Br)CH₂COOH
Boiling Pt: 320.2 °C (760 mm Hg)
Melting Pt: 104…106 °C
Flash Pt: 147.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004314
CAS Number: 18698-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR