2-(2-Bromophenyl)-1H-imidazole 97%

Supplier: Ambeed
Warning

A191258-1G A191258-5G A191258-100MG A191258-10G A191258-250MG A191258-25G
103816-358EA 33 USD
103816-358 103846-960 77084-788 77084-790 77236-900 77236-902 77236-904 77236-906
2-(2-Bromophenyl)-1H-imidazole 97%
2-(2-Bromophenyl)-1H-imidazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.069 g/mol
Boiling Pt: 397.3 °C (760 mm Hg)
Flash Pt: 194.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08668745
CAS Number: 162356-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR