N-(2-Bromoethyl)phthalimide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2-Bromoethyl)isoindoline-1,3-dione
A194659-100G
A194659-10G
A194659-500G
A194659-25G
A194659-5G
103847-292EA
7
USD
103847-292
76814-012
76814-014
76814-016
77302-428
77302-430
N-(2-Bromoethyl)phthalimide 98%
N-(2-Bromoethyl)phthalimide
Precaución: for Research Use Only
Formula:
C₁₀H₈BrNO₂ Boiling Pt: 318 °C (760 mm Hg) Melting Pt: 80…83 °C Flash Pt: 161.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005902 CAS Number: 574-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...