2-Bromo-5-fluorophenylacetonitrile 98%
Supplier: Ambeed
|
Warning
|
A120854-10G
A120854-250MG
A120854-25G
A120854-1G
A120854-5G
A120854-100G
103800-000EA
13
USD
103800-000
76781-740
76781-742
76781-744
77312-104
77312-106
77666-732
2-Bromo-5-fluorophenylacetonitrile 98%
2-Bromo-5-fluorophenylacetonitrile
Precaución: for Research Use Only
Formula:
C₈H₅BrFN Boiling Pt: 271.7 °C (760 mm Hg) Melting Pt: 70…71 °C Flash Pt: 118.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094439 CAS Number: 886761-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...