(2-Bromo-5-chlorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A100940-10G A100940-25G A100940-1G A100940-5G
103795-714EA 10 USD
103795-714 103821-402 77233-062 77233-064 77282-610 77296-920
(2-Bromo-5-chlorophenyl)acetic acid 97%
(2-Bromo-5-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
Melting Pt: 101…103 °C
Flash Pt: 162.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00153890
CAS Number: 81682-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR