2-Bromo-4,5-difluorophenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Bromo-4,5-difluorophenyl)acetic acid

A179419-250MG A179419-5G A179419-1G
103807-426EA 45 USD
103807-426 103845-772 76872-694 77083-224 77301-612
2-Bromo-4,5-difluorophenylacetic acid 97%
2-Bromo-4,5-difluorophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
MW: 251.027 g/mol
Boiling Pt: 313 °C (760 mm Hg)
Flash Pt: 143.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04973423
CAS Number: 883502-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR