2-Bromo-3-fluorophenylacetic acid 97%

Supplier: Ambeed
Warning

A512850-1G A512850-25G A512850-10G A512850-250MG
103828-420EA 27 USD
103828-420 103839-518 76843-504 76843-506 77673-362 77872-854
2-Bromo-3-fluorophenylacetic acid 97%
2-Bromo-3-fluorophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Boiling Pt: 312.2±27.0 °C (760 mm Hg)
Flash Pt: 142.6 °C
Storage Temperature: Ambient
MDL Number: MFCD09864672
CAS Number: 958454-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR