(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: (Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetic acid, syn-Thiazoximic Acid

A176035-100G A176035-10G A176035-500G A176035-25G A176035-5G
103845-424EA 6 USD
103845-424 77273-878 77273-880 77273-882 77301-418 77301-420
(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid 97%
(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid

Precaución: for Research Use Only

Formula: C₆H₇N₃O₃S
MW: 201.20 g/mol
Boiling Pt: 425.1 °C (760 mm Hg)
Melting Pt: 192 °C
Storage Temperature: Ambient
MDL Number: MFCD00010416
CAS Number: 65872-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR