2-(2,6-Dioxopiperidin-3-yl)-4-nitroisoindoline-1,3-dione 97%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A127237-250MG
A127237-5G
A127237-1G
A127237-25G
A127237-100G
103800-936EA
8
USD
103800-936
76983-502
76983-504
76983-506
77313-184
77611-098
77683-128
2-(2,6-Dioxopiperidin-3-yl)-4-nitroisoindoline-1,3-dione 97%, Ambeed.Inc
2-(2,6-Dioxopiperidin-3-yl)-4-nitroisoindoline-1,3-dione
Precaución: For research use only.
Formula:
C₁₃H₉N₃O₆ Boiling Pt: 603.4 °C (760 mm Hg) Flash Pt: 318.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD01748356 CAS Number: 19171-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...